Assay Details
| Gene Symbol: Ddx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110826.1 | NP_001104296.1 | 11 - 12 | 1534 | 138 | |
| NM_007841.4 | NP_031867.1 | 11 - 12 | 1477 | 138 | ||
| NM_181324.3 | NP_851841.2 | 11 - 12 | 1502 | 138 | ||
| GenBank mRNA | AF038995.1 | - | 10 - 11 | 1096 | 138 | |
| AK054144.1 | - | 11 - 12 | 1496 | 138 | ||
| AK148483.1 | - | 11 - 12 | 1499 | 138 | ||
| BC021452.1 | - | 10 - 11 | 1394 | 5043599 | 138 | |
More Information
Set Membership: |
|

Ddx6
Genomic Map