| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_001128086.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_001128086.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_001177396.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_001177396.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_001177397.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_001177397.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_001177398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_001177398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_181398.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
NM_181398.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008369.mm.1 |
TC1000001090.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_001128086.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_001128086.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_001177396.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_001177396.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_001177397.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_001177397.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_001177398.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_001177398.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_181398.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
NM_181398.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000008370.mm.1 |
TC1000001090.mm.1 |
MTA TC |