Assay Details
| Gene Symbol: Usp6nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080548.1 | NP_001074017.1 | 6 - 7 | 563 | 91 | |
| NM_181399.3 | NP_852064.2 | 5 - 6 | 402 | 91 | ||
| GenBank mRNA | AK084045.1 | - | 6 - 7 | 386 | 91 | |
| AK129037.1 | - | 6 - 7 | 415 | 91 | ||
| AK155073.1 | - | 6 - 7 | 564 | 91 | ||
| BC051184.1 | - | 5 - 6 | 405 | 91 | ||
| BQ938063.1 | - | 5 - 6 | 402 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp6nl
Genomic Map