Assay Details
| Gene Symbol: Parp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181402.3 | NP_852067.1 | 1 - 2 | 96 | 63 | |
| GenBank mRNA | AK133697.1 | - | 1 - 2 | 144 | 63 | |
| BC040269.1 | - | 1 - 2 | 96 | 4035469 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133697.1, BC040269.1, NM_181402.3 transcripts.
Set Membership: |
|

Parp11
Genomic Map