Assay Details
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028422.1 | NP_082698.1 | 3 - 4 | 257 | 139 | |
NM_181424.3 | NP_852089.2 | 4 - 5 | 351 | 139 | ||
GenBank mRNA | AK163448.1 | - | 4 - 5 | 362 | 139 | |
BC025144.1 | - | 4 - 5 | 350 | 4483063 | 139 | |
BY726952.1 | - | 4 - 5 | 362 | 139 | ||
CX210755.1 | - | 2 - 3 | 240 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC025144.1, NM_028422.1, NM_181424.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
