Assay Details
| Gene Symbol: Ahcyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171000.2 | NP_001164471.2 | 13 - 14 | 1659 | 65 | |
| NM_001171001.1 | NP_001164472.1 | 13 - 14 | 1379 | 65 | ||
| NM_021414.6 | NP_067389.5 | 13 - 14 | 1662 | 65 | ||
| GenBank mRNA | AK053527.1 | - | 13 - 14 | 1343 | 65 | |
| AK122382.1 | - | 11 - 12 | 1365 | 65 | ||
| AK161892.1 | - | 2 - 3 | 139 | 65 | ||
| BC037447.1 | - | 1 - 2 | 106 | 65 | ||
| BC051504.1 | - | 3 - 4 | 292 | 65 | ||
| BC059255.1 | - | 9 - 10 | 834 | 65 | ||
| BC079660.1 | - | 13 - 14 | 1572 | 6853550 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahcyl2
Genomic Map