Assay Details
| Gene Symbol: Ahcyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171000.2 | NP_001164471.2 | 15 - 16 | 1805 | 80 | |
| NM_001171001.1 | NP_001164472.1 | 15 - 16 | 1525 | 80 | ||
| NM_021414.6 | NP_067389.5 | 15 - 16 | 1808 | 80 | ||
| GenBank mRNA | AK014539.1 | - | 16 - 17 | 1759 | 80 | |
| AK053527.1 | - | 15 - 16 | 1489 | 80 | ||
| AK122382.1 | - | 13 - 14 | 1511 | 80 | ||
| AK161892.1 | - | 4 - 5 | 285 | 80 | ||
| BC037447.1 | - | 3 - 4 | 252 | 80 | ||
| BC051504.1 | - | 5 - 6 | 438 | 80 | ||
| BC059255.1 | - | 11 - 12 | 980 | 80 | ||
| BC079660.1 | - | 15 - 16 | 1718 | 6853550 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahcyl2
Genomic Map