| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100011214.mm.1 |
NM_001163354.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011214.mm.1 |
NM_001163354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011214.mm.1 |
NM_001163355.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011214.mm.1 |
NM_001163355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011214.mm.1 |
NM_021536.7.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011214.mm.1 |
NM_021536.7 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011214.mm.1 |
TC1100001198.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100011217.mm.1 |
NM_001163354.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011217.mm.1 |
NM_001163354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011217.mm.1 |
NM_001163355.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011217.mm.1 |
NM_001163355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011217.mm.1 |
NM_021536.7.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011217.mm.1 |
NM_021536.7 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011217.mm.1 |
TC1100001198.mm.1 |
MTA TC |