Assay Details
| Gene Symbol: Rhot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163354.1 | NP_001156826.1 | 3 - 4 | 405 | 154 | |
| NM_001163355.1 | NP_001156827.1 | 3 - 4 | 405 | 154 | ||
| NM_021536.7 | NP_067511.4 | 3 - 4 | 405 | 154 | ||
| GenBank mRNA | AK019059.1 | - | 3 - 4 | 365 | 154 | |
| AK028597.1 | - | 3 - 4 | 348 | 154 | ||
| AK031139.1 | - | 1 - 2 | 67 | 154 | ||
| AK032096.1 | - | 3 - 4 | 378 | 154 | ||
| AK041119.1 | - | 3 - 4 | 341 | 154 | ||
| AK054472.1 | - | 3 - 4 | 376 | 154 | ||
| AK078955.1 | - | 3 - 4 | 348 | 154 | ||
| AK138830.1 | - | 3 - 4 | 346 | 154 | ||
| AK141621.1 | - | 3 - 4 | 232 | 154 | ||
| AK147749.1 | - | 3 - 4 | 384 | 154 | ||
| AK147801.1 | - | 3 - 4 | 410 | 154 | ||
| AK167502.1 | - | 3 - 4 | 405 | 154 | ||
| AK169811.1 | - | 3 - 4 | 374 | 154 | ||
| AK191854.1 | - | 1 - 2 | 39 | 154 | ||
| BC046785.1 | - | 3 - 4 | 388 | 6416127 | 154 | |
| BC058350.1 | - | 3 - 4 | 320 | 6815832 | 154 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031139.1 transcripts.
Set Membership: |
|

Rhot1
Genomic Map