Assay Details
| Gene Symbol: Rhot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163354.1 | NP_001156826.1 | 7 - 8 | 668 | 132 | |
| NM_001163355.1 | NP_001156827.1 | 7 - 8 | 668 | 132 | ||
| NM_021536.7 | NP_067511.4 | 7 - 8 | 668 | 132 | ||
| GenBank mRNA | AK019059.1 | - | 7 - 8 | 628 | 132 | |
| AK028597.1 | - | 7 - 8 | 611 | 132 | ||
| AK031139.1 | - | 5 - 6 | 330 | 132 | ||
| AK032096.1 | - | 7 - 8 | 641 | 132 | ||
| AK041119.1 | - | 7 - 8 | 604 | 132 | ||
| AK053263.1 | - | 6 - 7 | 525 | 132 | ||
| AK054472.1 | - | 7 - 8 | 639 | 132 | ||
| AK078955.1 | - | 7 - 8 | 611 | 132 | ||
| AK138830.1 | - | 7 - 8 | 609 | 132 | ||
| AK141621.1 | - | 7 - 8 | 495 | 132 | ||
| AK147749.1 | - | 7 - 8 | 647 | 132 | ||
| AK147801.1 | - | 7 - 8 | 673 | 132 | ||
| AK167502.1 | - | 7 - 8 | 668 | 132 | ||
| AK169811.1 | - | 7 - 8 | 637 | 132 | ||
| AK191854.1 | - | 5 - 6 | 302 | 132 | ||
| BC046785.1 | - | 7 - 8 | 651 | 6416127 | 132 | |
| BC058350.1 | - | 7 - 8 | 583 | 6815832 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhot1
Genomic Map