Assay Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143776.1 | NP_001137248.1 | 6 - 7 | 755 | 78 | |
| NM_001143777.1 | NP_001137249.1 | 7 - 8 | 859 | 78 | ||
| NM_024244.4 | NP_077206.3 | 6 - 7 | 755 | 78 | ||
| GenBank mRNA | AK004797.1 | - | 6 - 7 | 739 | 78 | |
| AK021133.1 | - | 7 - 8 | 698 | 78 | ||
| AK036299.1 | - | 7 - 8 | 822 | 78 | ||
| AK041147.1 | - | 6 - 7 | 756 | 78 | ||
| AK044049.1 | - | 7 - 8 | 782 | 78 | ||
| AK082202.1 | - | 6 - 7 | 751 | 78 | ||
| AK220421.1 | - | 7 - 8 | 695 | 78 | ||
| BC129839.1 | - | 6 - 7 | 711 | 40060810 | 78 | |
| BC129840.1 | - | 6 - 7 | 711 | 40060811 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam13c
Genomic Map