Assay Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160208.1 | NP_001153680.1 | 1 - 2 | 41 | 73 | |
| NM_025505.4 | NP_079781.2 | 1 - 2 | 41 | 73 | ||
| GenBank mRNA | AK006544.1 | - | 1 - 2 | 45 | 73 | |
| AK029787.1 | - | 1 - 2 | 42 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006544.1, AK029787.1, NM_001160208.1, NM_025505.4 transcripts.
Set Membership: |
|

Blzf1
Genomic Map