Assay Details
| Gene Symbol: Gpatch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026367.4 | NP_080643.4 | 6 - 7 | 1270 | 88 | |
| GenBank mRNA | AK029990.1 | - | 6 - 7 | 1254 | 88 | |
| AK030026.1 | - | 6 - 7 | 1269 | 88 | ||
| AK083471.1 | - | 5 - 6 | 427 | 88 | ||
| BC054810.1 | - | 6 - 7 | 1242 | 6839419 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpatch2
Genomic Map