Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 24 - 25 | 2407 | 90 | |
| GenBank mRNA | AK087373.1 | - | 1 - 2 | 1306 | 90 | |
| AK087793.1 | - | 24 - 25 | 2440 | 90 | ||
| BC029746.1 | - | 24 - 25 | 2407 | 3494321 | 90 | |
| BC031900.1 | - | 25 - 26 | 2370 | 5364208 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo11
Genomic Map