Assay Details
| Gene Symbol: Ipo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029665.3 | NP_083941.2 | 25 - 26 | 2508 | 80 | |
| GenBank mRNA | AK087373.1 | - | 2 - 3 | 1408 | 80 | |
| AK087793.1 | - | 25 - 26 | 2541 | 80 | ||
| BC029746.1 | - | 25 - 26 | 2508 | 3494321 | 80 | |
| BC031900.1 | - | 26 - 27 | 2471 | 5364208 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ipo11
Genomic Map