Assay Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146068.4 | NP_666180.2 | 10 - 11 | 1671 | 87 | |
| GenBank mRNA | AK044836.1 | - | 10 - 11 | 1654 | 87 | |
| AK052832.1 | - | 10 - 11 | 1634 | 87 | ||
| AK075871.1 | - | 10 - 11 | 1636 | 87 | ||
| AK146053.1 | - | 10 - 11 | 1671 | 87 | ||
| AK170784.1 | - | 6 - 7 | 1303 | 87 | ||
| AK172902.1 | - | 10 - 11 | 1624 | 87 | ||
| BC046558.1 | - | 4 - 5 | 675 | 87 | ||
| BC069859.1 | - | 1 - 2 | 224 | 87 | ||
| BC085258.1 | - | 1 - 2 | 42 | 87 | ||
| BC089026.1 | - | 10 - 11 | 1608 | 30615298 | 87 | |
More Information
Set Membership: |
|

Spidr
Genomic Map