Assay Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146068.4 | NP_666180.2 | 5 - 6 | 660 | 101 | |
| GenBank mRNA | AK044836.1 | - | 5 - 6 | 643 | 101 | |
| AK052832.1 | - | 5 - 6 | 623 | 101 | ||
| AK075871.1 | - | 5 - 6 | 625 | 101 | ||
| AK146053.1 | - | 5 - 6 | 660 | 101 | ||
| AK172902.1 | - | 5 - 6 | 613 | 101 | ||
| BC089026.1 | - | 5 - 6 | 597 | 30615298 | 101 | |
More Information
Set Membership: |
|

Spidr
Genomic Map