Assay Details
| Gene Symbol: Fam160a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172682.3 | NP_766270.2 | 6 - 7 | 1380 | 62 | |
| GenBank mRNA | AK036881.1 | - | 3 - 4 | 366 | 62 | |
| BC026906.1 | - | 2 - 3 | 189 | 62 | ||
| BC043298.1 | - | 3 - 4 | 332 | 62 | ||
| BC094511.1 | - | 4 - 5 | 591 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam160a1
Genomic Map