Assay Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110496.1 | NP_001103966.1 | 10 - 11 | 1039 | 77 | |
| NM_001110497.1 | NP_001103967.1 | 10 - 11 | 1039 | 77 | ||
| NM_173734.3 | NP_776095.2 | 10 - 11 | 1039 | 77 | ||
| GenBank mRNA | AK044591.1 | - | 10 - 11 | 993 | 77 | |
| AK086738.1 | - | 10 - 11 | 989 | 77 | ||
| AK088559.1 | - | 10 - 11 | 1021 | 77 | ||
| AK134268.1 | - | 10 - 11 | 1002 | 77 | ||
| BC004047.1 | - | 10 - 11 | 988 | 77 | ||
| BC027354.1 | - | 10 - 11 | 1012 | 4010477 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem87a
Genomic Map