Assay Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110496.1 | NP_001103966.1 | 13 - 14 | 1309 | 77 | |
| NM_001110497.1 | NP_001103967.1 | 13 - 14 | 1309 | 77 | ||
| NM_173734.3 | NP_776095.2 | 13 - 14 | 1312 | 77 | ||
| GenBank mRNA | AK044591.1 | - | 13 - 14 | 1263 | 77 | |
| AK086738.1 | - | 13 - 14 | 1259 | 77 | ||
| AK088559.1 | - | 13 - 14 | 1294 | 77 | ||
| AK134268.1 | - | 13 - 14 | 1275 | 77 | ||
| BC004047.1 | - | 13 - 14 | 1258 | 77 | ||
| BC024676.1 | - | 3 - 4 | 260 | 77 | ||
| BC027354.1 | - | 13 - 14 | 1285 | 4010477 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem87a
Genomic Map