Assay Details
| Gene Symbol: Hmces | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173737.2 | NP_776098.1 | 3 - 4 | 384 | 91 | |
| GenBank mRNA | AK010550.1 | - | 1 - 2 | 152 | 91 | |
| AK018400.1 | - | 2 - 3 | 534 | 91 | ||
| BC024401.1 | - | 3 - 4 | 384 | 5026919 | 91 | |
| BC064070.1 | - | 3 - 4 | 367 | 3371288 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmces
Genomic Map