Assay Details
| Gene Symbol: Hmces | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173737.2 | NP_776098.1 | 4 - 5 | 514 | 80 | |
| GenBank mRNA | AK010550.1 | - | 2 - 3 | 282 | 80 | |
| AK018400.1 | - | 3 - 4 | 664 | 80 | ||
| BC024401.1 | - | 4 - 5 | 514 | 5026919 | 80 | |
| BC064070.1 | - | 4 - 5 | 497 | 3371288 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618849 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100963 | BC064070.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100963 | AK010550.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100963 | AK018400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100963 | NM_173737.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100963 | BC024401.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305225 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371665 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166491 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44234 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118497 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160765 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100963 | BC064070.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100963 | AK010550.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100963 | AK018400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100963 | NM_173737.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100963 | BC024401.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618851 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118497 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160765 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05972748 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05972784 | - | - |
| Whole Mouse Genome G4122A | A_51_P298323 | BC064070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P298323 | AK010550.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P298323 | AK018400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P298323 | NM_173737.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P298323 | AK215958.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504400 | BC064070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504400 | AK010550.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504400 | AK018400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504400 | NM_173737.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504400 | AK215958.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138545 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118497 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160765 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05545515 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05128841 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05128843 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05545519 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05128848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24618847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05128847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05415590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05790135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05539215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05116228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05539209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05844779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05128843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24618871 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539856 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539855 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539854 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539853 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423786_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539850 | - | - |
| Mouse Genome 430 2.0 Array | 1423786_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423786_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232210_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232210_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600008012.mm.1 | TC0600001020.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600008012.mm.1 | NM_173737.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600008012.mm.1 | NM_173737.2 | Refseq |
| Mouse Expression Set 430 | 1423786_at | - | - |
| Mouse Expression Set 430 | 1423786_PM_at | - | - |
| Murine Genome U74v2 Set | 160684_PM_at | - | - |
| Murine Genome U74v2 Set | 160684_at | - | - |
| Murine Genome U74v2 Set | 168394_f_at | - | - |
| Murine Genome U74v2 Set | 168394_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423786_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423786_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423786_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423786_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 645588 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6590128 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590128 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2604264 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2604264 | - | - |
| mouse-6 v1.1 (Illumina) | scl29779.8.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590128 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29779.8.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590128 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2604263 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2604263 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29779.8.1_30-S | - | - |

Hmces
Genomic Map