Assay Details
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291049.1 | NP_001277978.1 | 6 - 7 | 1210 | 74 | |
NM_173754.4 | NP_776115.2 | 6 - 7 | 1210 | 74 | ||
GenBank mRNA | AK042217.1 | - | 6 - 7 | 1211 | 74 | |
AK083271.1 | - | 6 - 7 | 1048 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
