Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293560.1 | NP_001280489.1 | 4 - 5 | 439 | 125 | |
| NM_173763.4 | NP_776124.1 | 4 - 5 | 443 | 125 | ||
| GenBank mRNA | AK049569.1 | - | 4 - 5 | 412 | 125 | |
| AK145623.1 | - | 4 - 5 | 437 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl2
Genomic Map