Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293560.1 | NP_001280489.1 | 5 - 6 | 585 | 142 | |
| NM_173763.4 | NP_776124.1 | 5 - 6 | 589 | 142 | ||
| GenBank mRNA | AF363737.1 | - | 5 - 6 | 559 | 142 | |
| AK049569.1 | - | 5 - 6 | 558 | 142 | ||
| AK145623.1 | - | 5 - 6 | 583 | 142 | ||
| BC037135.1 | - | 1 - 2 | 166 | 142 | ||
| BC085287.1 | - | 6 - 7 | 682 | 142 | ||
| BC131942.1 | - | 4 - 5 | 374 | 40130219 | 142 | |
| BC132615.1 | - | 4 - 5 | 374 | 40130892 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl2
Genomic Map