Assay Details
| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173775.3 | NP_776136.2 | 13 - 14 | 1446 | 74 | |
| GenBank mRNA | AK048790.1 | - | 5 - 6 | 2041 | 74 | |
| BC047211.1 | - | 11 - 12 | 1128 | 6392183 | 74 | |
| BC079632.1 | - | 1 - 2 | 174 | 5693354 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc37
Genomic Map