Assay Details
| Gene Symbol: Rab6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173781.4 | NP_776142.1 | 5 - 6 | 480 | 76 | |
| GenBank mRNA | AB232597.1 | - | 5 - 6 | 397 | 76 | |
| AK035893.1 | - | 5 - 6 | 439 | 76 | ||
| BC059903.1 | - | 1 - 2 | 63 | 76 | ||
| BC060618.1 | - | 5 - 6 | 480 | 6406134 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab6b
Genomic Map