Assay Details
| Gene Symbol: Ppm1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110218.1 | NP_001103688.1 | 2 - 3 | 834 | 60 | |
| NM_176919.4 | NP_795893.2 | 2 - 3 | 834 | 60 | ||
| GenBank mRNA | AK029461.1 | - | 2 - 3 | 396 | 60 | |
| AK040194.1 | - | 2 - 3 | 578 | 60 | ||
| AK040207.1 | - | 2 - 3 | 819 | 60 | ||
| AK043121.1 | - | 2 - 3 | 835 | 60 | ||
| AK051434.1 | - | 2 - 3 | 792 | 60 | ||
| AK134804.1 | - | 2 - 3 | 814 | 60 | ||
| AK150309.1 | - | 2 - 3 | 396 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1h
Genomic Map