Assay Details
| Gene Symbol: Zcchc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138590.4 | NP_613056.2 | 4 - 5 | 923 | 152 | |
| GenBank mRNA | AK029808.1 | - | 4 - 5 | 930 | 152 | |
| AK032353.1 | - | 4 - 5 | 895 | 152 | ||
| AK039158.1 | - | 4 - 5 | 844 | 152 | ||
| AK043238.1 | - | 4 - 5 | 836 | 152 | ||
| AK080785.1 | - | 5 - 6 | 1012 | 152 | ||
| AK157728.1 | - | 4 - 5 | 826 | 152 | ||
| BC050200.1 | - | 3 - 4 | 798 | 152 | ||
| BC117020.1 | - | 4 - 5 | 826 | 40126339 | 152 | |
| BC117022.1 | - | 4 - 5 | 826 | 40126341 | 152 | |
| BC144774.1 | - | 4 - 5 | 830 | 9053322 | 152 | |
| CF732773.1 | - | 3 - 4 | 514 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zcchc7
Genomic Map