Assay Details
| Gene Symbol: Oacyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177028.3 | NP_796002.1 | 9 - 10 | 1298 | 65 | |
| GenBank mRNA | AK030604.1 | - | 9 - 10 | 1298 | 65 | |
| AK030615.1 | - | 7 - 8 | 1078 | 65 | ||
| BC141265.1 | - | 9 - 10 | 1295 | 9055877 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oacyl
Genomic Map