Assay Details
| Gene Symbol: Apba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177034.3 | NP_796008.2 | 7 - 8 | 1901 | 88 | |
| GenBank mRNA | AK032261.1 | - | 6 - 7 | 665 | 88 | |
| AK147583.1 | - | 7 - 8 | 1891 | 88 | ||
| AK147623.1 | - | 7 - 8 | 1853 | 88 | ||
| BC141181.1 | - | 7 - 8 | 1719 | 9055793 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apba1
Genomic Map