Assay Details
| Gene Symbol: Ceacam19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177036.5 | NP_796010.1 | 5 - 6 | 864 | 78 | |
| GenBank mRNA | AK079290.1 | - | 5 - 6 | 865 | 78 | |
| AK163794.1 | - | 5 - 6 | 852 | 78 | ||
| BC107227.1 | - | 5 - 6 | 792 | 40051960 | 78 | |
| BC107228.2 | - | 5 - 6 | 792 | 40051962 | 78 | |
| BC109333.3 | - | 5 - 6 | 792 | 40051723 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967727 | AK079290.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1967727 | NM_177036.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110021 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161535 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153222 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967727 | AK079290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1967727 | NM_177036.5 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24995732 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161535 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153222 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06136389 | - | - |
| Whole Mouse Genome G4122A | A_51_P467238 | AK079290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P467238 | AK163794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P467238 | AK047942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P467238 | NM_177036.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504237 | BC107228.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504237 | BC109333.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504237 | AK079290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504237 | AK163794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504237 | BC107227.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121125 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153222 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24995682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24995755 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10560706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560708 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560704 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560702 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022184.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022184.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022184.mm.1 | NM_177036.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022187.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022187.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022187.mm.1 | NM_177036.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022188.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022188.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022188.mm.1 | NM_177036.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022185.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022185.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022185.mm.1 | NM_177036.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022186.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022186.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022186.mm.1 | NM_177036.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022189.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022189.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022189.mm.1 | NM_177036.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022190.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022190.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022190.mm.1 | NM_177036.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022192.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022192.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022192.mm.1 | NM_177036.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022191.mm.1 | TC0700002506.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022191.mm.1 | NM_177036.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022191.mm.1 | NM_177036.5 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2630309 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2120138 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2120138 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2630309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002120138 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775467 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239323 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775467 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239323 | - | - |
| mouse-6 v1.1 (Illumina) | scl00319930.2_238-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0319930.1_260-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2120138 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0319930.1_260-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00319930.2_238-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120138 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630309 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239323 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239323 | - | - |

Ceacam19
Genomic Map