Assay Details
| Gene Symbol: Trappc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029491.2 | NP_083767.2 | 10 - 11 | 1497 | 70 | |
| NM_177038.3 | NP_796012.2 | 10 - 11 | 1497 | 70 | ||
| GenBank mRNA | AK083747.1 | - | 10 - 11 | 1497 | 70 | |
| AK173085.1 | - | 3 - 4 | 274 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trappc8
Genomic Map