Assay Details
| Gene Symbol: Rad9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042331.1 | - | 1 | 187 | 70 | |
| AK081488.1 | - | 1 | 153 | 70 | ||
| AK157179.1 | - | 1 | 181 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rad9b
Genomic Map