Assay Details
| Gene Symbol: Lrig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177152.5 | NP_796126.4 | 5 - 6 | 916 | 89 | |
| GenBank mRNA | AK035166.1 | - | 5 - 6 | 918 | 89 | |
| AK137007.1 | - | 5 - 6 | 903 | 89 | ||
| AY505341.1 | - | 5 - 6 | 859 | 89 | ||
| BC065142.1 | - | 5 - 6 | 882 | 30610561 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrig3
Genomic Map