Assay Details
| Gene Symbol: Klri2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177155.4 | NP_796129.2 | 1 - 2 | 287 | 80 | |
| GenBank mRNA | AK040915.1 | - | 1 - 2 | 274 | 80 | |
| AK041213.1 | - | 1 - 2 | 290 | 80 | ||
| AY324873.1 | - | 1 - 2 | 240 | 80 | ||
| BC132406.1 | - | 1 - 2 | 267 | 40130683 | 80 | |
| BC132408.1 | - | 1 - 2 | 267 | 40130685 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119377 | AK040915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2119377 | NM_177155.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296037 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342195 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68396 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164230 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142017 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119377 | AK040915.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2119377 | NM_177155.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24845262 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164230 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142017 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06068701 | - | - |
| Whole Mouse Genome G4122A | A_52_P340094 | AK040915.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P340094 | NM_177155.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P387294 | BC132406.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P387294 | BC132408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P387294 | AK040915.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P387294 | AK041213.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P387294 | NM_177155.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117274 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164230 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142017 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05403409 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05403403 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05095145 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05403406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05095146 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05095144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24845157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05813399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05813393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05754047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05114324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P04998830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05095148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05403407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05170075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05403402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24845233 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548427 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548422 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025382.mm.1 | TC0600003247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025382.mm.1 | NM_177155.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025382.mm.1 | NM_177155.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025383.mm.1 | TC0600003247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025383.mm.1 | NM_177155.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025383.mm.1 | NM_177155.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025380.mm.1 | TC0600003247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025380.mm.1 | NM_177155.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025380.mm.1 | NM_177155.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025378.mm.1 | TC0600003247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025378.mm.1 | NM_177155.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025378.mm.1 | NM_177155.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025381.mm.1 | TC0600003247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025381.mm.1 | NM_177155.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025381.mm.1 | NM_177155.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025379.mm.1 | TC0600003247.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025379.mm.1 | NM_177155.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025379.mm.1 | NM_177155.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 496003 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5420068 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420068 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2651722 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2651722 | - | - |
| mouse-6 v1.1 (Illumina) | scl28330.6.1_242-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420068 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28330.6.1_242-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420068 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2651722 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2651722 | - | - |

Klri2
Genomic Map