Assay Details
| Gene Symbol: P4ha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177161.4 | NP_796135.3 | 7 - 8 | 1116 | 128 | |
| GenBank mRNA | AK086468.1 | - | 5 - 6 | 704 | 128 | |
| AY313449.1 | - | 7 - 8 | 1116 | 128 | ||
| BC082538.1 | - | 7 - 8 | 1158 | 5701368 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

P4ha3
Genomic Map