Assay Details
| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177162.4 | NP_796136.2 | 9 - 10 | 1192 | 123 | |
| GenBank mRNA | AK036981.1 | - | 9 - 10 | 1193 | 123 | |
| AK037029.1 | - | 5 - 6 | 690 | 123 | ||
| BC118611.1 | - | 9 - 10 | 1170 | 40107184 | 123 | |
| BC118660.1 | - | 9 - 10 | 1169 | 40107187 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110914 | AK036981.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110914 | AK037029.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110914 | NM_177162.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892868 | XM_001480657.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892868 | XM_001480660.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892868 | XM_001480686.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892868 | XM_001480689.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251095 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335439 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270395 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670628 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135815 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104710 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157060 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110914 | AK036981.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110914 | AK037029.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110914 | NM_177162.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P151133 | AK036981.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157060 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104710 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05643124 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01152828 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05643074 | - | - |
| Whole Mouse Genome G4122A | A_51_P151133 | AK036981.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110914 | AK036981.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110914 | AK037029.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110914 | NM_177162.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157060 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23864119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23863965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04138854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04823662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04832474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04223689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04918184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04138853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04466408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23864118 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10530989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530988 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530997 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530993 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10530996 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438337_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10530986 | - | - |
| Mouse Genome 430 2.0 Array | 1438337_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438337_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320454_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320454_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500023672.mm.1 | TC0500002683.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023672.mm.1 | NM_177162.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023672.mm.1 | NM_177162.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023671.mm.1 | TC0500002683.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023671.mm.1 | NM_177162.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023671.mm.1 | NM_177162.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023673.mm.1 | TC0500002683.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023673.mm.1 | NM_177162.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023673.mm.1 | NM_177162.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023675.mm.1 | TC0500002683.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023675.mm.1 | NM_177162.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023675.mm.1 | NM_177162.4 | Refseq |
| Mouse Expression Set 430 | 1438337_PM_x_at | - | - |
| Mouse Expression Set 430 | 1438337_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438337_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438337_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 419081 | - | - |
| ABI Mouse Genome Survey Microarray | 763428 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 70039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 70039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259406 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259406 | - | - |
| mouse-6 v1.1 (Illumina) | scl26428.11.1_197-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26428.11.1_197-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1259406 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259406 | - | - |

Tmprss11g
Genomic Map