Assay Details
Gene Symbol: Bhlha9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177182.4 | NP_796156.3 | 1 | 407 | 76 | |
GenBank mRNA | AK043927.1 | - | 1 | 152 | 76 | |
BC048728.1 | - | 1 | 407 | 6510150 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933408 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933358 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933361 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933405 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933360 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933409 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933406 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933359 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985288 | BC048728.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985288 | NM_177182.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348690 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107805 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985288 | BC048728.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985288 | NM_177182.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933409 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933366 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933400 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933408 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933411 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933397 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933378 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933367 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933410 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933394 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933373 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933386 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933381 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933371 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933382 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933364 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933374 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933395 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933392 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933403 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933376 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933402 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933368 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933390 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27933398 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107805 | - | - |
Whole Mouse Genome G4122A | A_51_P107463 | BC048728.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107463 | AK043927.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P107463 | NM_177182.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123929 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107805 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09099286 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09099272 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09531867 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09531868 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09721567 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933361 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933409 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933360 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933405 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933358 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933406 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933359 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27933408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27933351 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08986141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09009144 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933390 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09078000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09331167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09637292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09030907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09141997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933387 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09517844 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09099286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09461284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933364 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27933373 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10378786 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1446501_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10378785 | - | - |
Mouse Genome 430 2.0 Array | 1446501_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1446501_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320522_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320522_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100010025.mm.1 | NM_177182.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100010025.mm.1 | NM_177182.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100010025.mm.1 | TC1100001105.mm.1 | MTA TC |
Mouse Expression Set 430 | 1446501_at | - | - |
Mouse Expression Set 430 | 1446501_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446501_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446501_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 462772 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 580670 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 580670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000580670 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2656915 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2656915 | - | - |
mouse-6 v1.1 (Illumina) | scl41244.1.256_37-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 580670 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl41244.1.256_37-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580670 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2656915 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2656915 | - | - |
