Assay Details
| Gene Symbol: Dennd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177192.3 | NP_796166.2 | 9 - 10 | 2611 | 96 | |
| GenBank mRNA | AK038858.1 | - | 9 - 10 | 2512 | 96 | |
| AK050728.1 | - | 9 - 10 | 2469 | 96 | ||
| AK135015.1 | - | 9 - 10 | 2496 | 96 | ||
| BC132199.1 | - | 9 - 10 | 2316 | 40130476 | 96 | |
| BC132535.1 | - | 9 - 10 | 2316 | 40130812 | 96 | |
| BC145128.1 | - | 9 - 10 | 2316 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd5b
Genomic Map