Assay Details
| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 3 - 4 | 206 | 103 | |
| GenBank mRNA | AK076708.1 | - | 3 - 4 | 207 | 103 | |
| AK078567.1 | - | 3 - 4 | 175 | 103 | ||
| EF377342.1 | - | 3 - 4 | 195 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp8b5
Genomic Map