Assay Details
| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290138.1 | NP_001277067.1 | 5 - 6 | 1088 | 87 | |
| NM_001290141.1 | NP_001277070.1 | 5 - 6 | 563 | 87 | ||
| NM_177203.3 | NP_796177.2 | 5 - 6 | 467 | 87 | ||
| NR_110776.1 | 5 - 6 | 467 | 87 | |||
| NR_110780.1 | 4 - 5 | 332 | 87 | |||
| NR_110781.1 | 5 - 6 | 366 | 87 | |||
| GenBank mRNA | AK042903.1 | - | 5 - 6 | 1090 | 87 | |
| BC150827.1 | - | 4 - 5 | 285 | 87 | ||
| BC171976.1 | - | 4 - 5 | 285 | 87 | ||
| BC171978.1 | - | 5 - 6 | 319 | 87 | ||
| BC171981.1 | - | 4 - 5 | 285 | 87 | ||
| BU962217.1 | - | 469 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc169
Genomic Map