Assay Details
| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290138.1 | NP_001277067.1 | 6 - 7 | 1147 | 74 | |
| NM_001290141.1 | NP_001277070.1 | 6 - 7 | 622 | 74 | ||
| NM_177203.3 | NP_796177.2 | 6 - 7 | 526 | 74 | ||
| NR_110776.1 | 6 - 7 | 526 | 74 | |||
| GenBank mRNA | AK042903.1 | - | 6 - 7 | 1149 | 74 | |
| BC150827.1 | - | 5 - 6 | 344 | 74 | ||
| BC171981.1 | - | 5 - 6 | 344 | 74 | ||
| BU962217.1 | - | 528 | 74 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc169
Genomic Map