Assay Details
| Gene Symbol: Abca15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177213.3 | NP_796187.2 | 7 - 8 | 1141 | 112 | |
| GenBank mRNA | AK029712.1 | - | 7 - 8 | 1141 | 112 | |
| AY243471.1 | - | 7 - 8 | 1141 | 112 | ||
| BC141350.1 | - | 7 - 8 | 1106 | 9055962 | 112 | |
| BC141351.1 | - | 7 - 8 | 1106 | 9055963 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca15
Genomic Map