Assay Details
| Gene Symbol: Cyb5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205227.1 | NP_001192156.1 | 8 - 9 | 836 | 95 | |
| NM_177216.4 | NP_796190.1 | 8 - 9 | 788 | 95 | ||
| GenBank mRNA | AK085272.1 | - | 8 - 9 | 753 | 95 | |
| AK147952.1 | - | 8 - 9 | 837 | 95 | ||
| BC107238.1 | - | 8 - 9 | 732 | 40052160 | 95 | |
| BC107239.1 | - | 8 - 9 | 732 | 40052161 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090214 | AK147952.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090214 | NM_001205227.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090214 | NM_177216.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090214 | BC100346.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090214 | NM_001163557.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912547 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655012 | AK076142.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655012 | AK081673.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655012 | AK085104.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655012 | AK173319.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655012 | BC062943.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862471 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85969 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125482 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090214 | AK147952.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090214 | NM_001205227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090214 | NM_177216.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090214 | BC100346.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090214 | NM_001163557.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25401502 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125482 | - | - |
| Whole Mouse Genome G4122A | A_51_P175858 | AK147952.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175858 | BC107238.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175858 | BC107239.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175858 | NM_001205227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175858 | NM_177216.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119091 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125482 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06574284 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06574283 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06574285 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06574271 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05913099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25401502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06324419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06643457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06118705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06574283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05929856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06118703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06653309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06643468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05994658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06643459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06574277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25401521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05913099 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566633 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459448_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566632 | - | - |
| Mouse Genome 430 2.0 Array | 1459448_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459448_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320635_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320635_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700033487.mm.1 | NM_001205227.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033487.mm.1 | NM_001205227.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033487.mm.1 | NM_177216.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033487.mm.1 | NM_177216.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033487.mm.1 | TC0700004038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700033488.mm.1 | NM_001205227.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033488.mm.1 | NM_001205227.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033488.mm.1 | NM_177216.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033488.mm.1 | NM_177216.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033488.mm.1 | TC0700004038.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1459448_at | - | - |
| Mouse Expression Set 430 | 1459448_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459448_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459448_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 338095 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 780441 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106110600 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 780441 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106110600 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000780441 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2655465 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2655465 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2468855 | - | - |
| mouse-6 v1.1 (Illumina) | scl30855.9.1_104-S | - | - |
| mouse-6 v1.1 (Illumina) | scl9362.1.1_255-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 780441 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106110600 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30855.9.1_104-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106110600 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780441 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2655465 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2655465 | - | - |

Cyb5r2
Genomic Map