Assay Details
| Gene Symbol: Hnrnpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177301.5 | NP_796275.3 | 5 - 6 | 819 | 89 | |
| GenBank mRNA | AK044748.1 | - | 5 - 6 | 595 | 89 | |
| AK085906.1 | - | 5 - 6 | 770 | 89 | ||
| AK144684.1 | - | 5 - 6 | 773 | 89 | ||
| BC027206.1 | - | 5 - 6 | 796 | 4218471 | 89 | |
| BC099683.1 | - | 5 - 6 | 814 | 6530863 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpl
Genomic Map