Assay Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159389.1 | NP_001152861.1 | 13 - 14 | 1487 | 77 | |
| NM_177306.3 | NP_796280.1 | 10 - 11 | 1118 | 77 | ||
| GenBank mRNA | AK029809.1 | - | 10 - 11 | 1119 | 77 | |
| AK049380.1 | - | 13 - 14 | 1467 | 77 | ||
| BC132104.1 | - | 8 - 9 | 801 | 40130381 | 77 | |
| BC132294.1 | - | 8 - 9 | 801 | 40130571 | 77 | |
| FJ804414.1 | - | 12 - 13 | 1335 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx6
Genomic Map