Assay Details
| Gene Symbol: Serac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111017.1 | NP_001104487.1 | 7 - 8 | 732 | 100 | |
| NM_177311.4 | NP_796285.2 | 6 - 7 | 642 | 100 | ||
| GenBank mRNA | AK019677.1 | - | 6 - 7 | 631 | 100 | |
| AK154605.1 | - | 7 - 8 | 683 | 100 | ||
| AK155571.1 | - | 7 - 8 | 735 | 100 | ||
| AK162246.1 | - | 7 - 8 | 667 | 100 | ||
| AK170498.1 | - | 7 - 8 | 683 | 100 | ||
| BC119594.1 | - | 7 - 8 | 721 | 40059066 | 100 | |
| BC119595.1 | - | 7 - 8 | 721 | 40059071 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Serac1
Genomic Map