Assay Details
| Gene Symbol: Dars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177445.5 | NP_803228.2 | 8 - 9 | 150 | ||
| GenBank mRNA | AK051206.1 | - | 1 - 2 | 150 | ||
| AK077533.1 | - | 8 - 9 | 150 | |||
| AK146350.1 | - | 8 - 9 | 819 | 150 | ||
| AK151027.1 | - | 8 - 9 | 150 | |||
| AK151897.1 | - | 8 - 9 | 150 | |||
| AK152304.1 | - | 8 - 9 | 150 | |||
| AK152562.1 | - | 8 - 9 | 150 | |||
| AK153256.1 | - | 8 - 9 | 150 | |||
| AK153299.1 | - | 8 - 9 | 150 | |||
| AK166009.1 | - | 8 - 9 | 150 | |||
| AK169247.1 | - | 8 - 9 | 150 | |||
| AK169716.1 | - | 8 - 9 | 150 | |||
| BC008638.1 | - | 8 - 9 | 773 | 3586278 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dars
Genomic Map