Assay Details
| Gene Symbol: Mogat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177448.4 | NP_803231.1 | 3 - 4 | 517 | 90 | |
| GenBank mRNA | AK133854.1 | - | 3 - 4 | 525 | 90 | |
| AY157609.1 | - | 3 - 4 | 474 | 90 | ||
| BC052831.1 | - | 3 - 4 | 521 | 30041797 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mogat2
Genomic Map